Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
HPLC Analysis of Miconazole Enantiomers on Astec® CYCLOBOND® I 2000 RSP
HPLC Analysis of Miconazole Enantiomers on Astec® CYCLOBOND® I 2000 RSP
Conditions
column CYCLOBOND I 2000 RSP, 25 cm x 4.6 mm I.D., 5 µm particles (20324AST)
column temp. ambient
mobile phase [A] 1% triethylamine acetate, pH 4.1; [B] acetonitrile; (80:20, A:B)
flow rate 2 mL/min
sample 5 mg/mL in methanol
injection 5 µL
detector UV, 230 nm
Description
Categories Analytical Chromatography, Chiral Applications, Pharmaceutical (small molecule), Antibiotics, Antimicrobials, All Pharmaceutical Applications, Antifungals
Featured Industry Pharmaceutical (small molecule)
Legal Information Astec is a registered trademark of Sigma-Aldrich Co. LLC, CYCLOBOND is a registered trademark of Sigma-Aldrich Co. LLC