HPLC Analysis of Mixture of Anisole and Related Compounds on Chromolith(R) Performance Diol 100-4.6mm
HPLC Analysis of Mixture of Anisole and Related Compounds on Chromolith® Performance Diol 100-4.6mm
Conditions
column Chromolith® Performance DIOL, 100 x 4.6 mm (1.53172.0001)
column temp. 23 °C
mobile phase [A] n-Heptane ; [B] 1,4-Dioxane; (95:5, A:B)
flow rate 1.3 mL/min
pressure 203 psi (14 Bar)
sample Anisole 390μg/ml, 3-Nitroanisole 70μg/ml, 4-Nitroanisole 260μg/ml, 2-Nitroanisole 180μg/ml (Diluent: Mobile phase)
injection 5 μL
detector 254 nm
Description
Featured Industry Vitamins, Nutraceuticals, and Natural Products
Legal Information Chromolith is a registered trademark of Merck KGaA, Darmstadt, Germany