Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
HPLC Analysis of Mouse Monoclonal Antibodies (MAb) on TSKgel® Ether-5PW
HPLC Analysis of Mouse Monoclonal Antibodies (MAb) on TSKgel® Ether-5PW
Conditions
column TSKgel Ether-5PW, 7.5 cm x 8 mm I.D., 10 µm particles (814014)
mobile phase [A] 0.05 M sodium phosphate, pH 7.0, 2 M ammonium sulfate, 1 M glycine; [B] 0.05 M sodium phosphate, pH 7.0, 1 M glycine
gradient linear gradient as shown on the figure
flow rate 1 mL/min
detector UV, 280 nm
Description
Categories Analytical Chromatography, Antibodies
Featured Industry Life Science and Biopharma
Legal Information TSKgel is a registered trademark of Tosoh Corporation
Other Notes Data provided by Tosoh Bioscience LLC.