Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
HPLC Analysis of mPEG and mPEG-Peptide on Zenix® and Zenix®-C SEC
HPLC Analysis of mPEG and mPEG-Peptide on Zenix® and Zenix®-C SEC
Conditions
column Zenix-C SEC-300, 30 cm x 7.8 mm, I.D., 3 µm particles (Z777092)
column Zenix SEC-300, 30 cm x 7.8 mm I.D., 3 µm particles (Z777033)
column temp. 25 °C
mobile phase 150 mM sodium phosphate buffer, pH 7.0
flow rate 1.0 mL/min
sample 4kD peptide and 20kD mPEG-MAL(methoxy-PEG-Maleimide) (6 mg/mL)
injection 20 µL
detector UV, 214 nm
Description
Categories Analytical Chromatography, Peptides
Featured Industry Life Science and Biopharma
Pharmaceutical (small molecule)
Legal Information Zenix is a registered trademark of Sepax Technologies