Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
HPLC Analysis of N-Linked Glycans on BIOshell™ Glycan
HPLC Analysis of N-Linked Glycans on BIOshell™ Glycan
Conditions
column BIOshell™ Glycan HPLC Column (50993-U)
column temp. 50 °C
mobile phase (A) Water with 100 mM ammonium formate, pH 4.5; (B) Acetonitrile
gradient min, %B, Flow (mL/min): 0-74-0.4; 15-71-0.4; 25-55-0.4; 26-45-0.2; 30-45-0.2; 31-75-0.4; 35-75-0.4.
pressure 1130 psi (78 bar)
sample in 70% mobile phase B
injection 3 µL
detector Fluorescence (λex = 260 nm, λem = 430 nm)
Description
Analysis Note Separation pf a mix of 2-AB labled N-linked glycans released from human IgG was optimized on BIOshell Glycan HPLC Columns. Baseline resolutions for all of the individual glycans were observed in both mixes, except for the positional isomer pair G1F and G1F’ which were partially resolved. 
Categories Analytical Chromatography, Proteins
Featured Industry Life Science and Biopharma
Legal Information BIOshell is a trademark of Sigma-Aldrich Co. LLC