Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
HPLC Analysis of Nucleosides on Ascentis® Si (Silica)
HPLC Analysis of Nucleosides on Ascentis® Si (Silica)
Conditions
column Ascentis Si, 10 cm x 2.1 mm I.D., 5 µm particles (581508-U)
column temp. 35 °C
mobile phase [A] 150 mM ammonium formate (pH 3.0, with formic acid): [B] acetonitrile; (10:90, A:B)
flow rate 0.2 mL/min
sample 100 µg/mL in mobile phase
injection 1 µL
detector UV, 270 nm
Description
Categories Analytical Chromatography, Nucleic Acids, Nucleosides