Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
HPLC Analysis of Organic acids on Chromolith(R) Performance 100 - 4.6 mm
HPLC Analysis of Organic acids on Chromolith® Performance 100 - 4.6 mm
Conditions
column Chromolith® Performance, 100 x 4.6 mm (1.52028.0001)
column temp. ambient
mobile phase [A] acetonitrile; [B] na-phosphatbuffer 20mM pH=2.5
gradient Initial composition: 0-0.5min 100%B to 82%B, hold until 5min, 5-6min 82%B to 100%B, hold until 8.5min, 8.5-12min 100%B to 55%B, hold until 16min
flow rate 1 mL/min
pressure 940 psi (56-65 bar)
sample Malic acid (0,5mg/ml), Fumaric acid (0,15mg/ml), Glutaric acid (0,7mg/ml), 3,4-Dihydroxy-cinnamic acid (0,1mg/ml), 4-Hydroxy-cinnamic acid (0,1mg/ml), Sorbic acid (0,75mg/ml), Benzoic acid (0,2mg/ml), 2-Hydroxybenzoic acid (0,5mg/ml), Cinnamic acid (0,2mg/ml)
injection 5 μL
detector 220 nm cell: 11 μL
Description
Featured Industry Food and Beverages
Legal Information Chromolith is a registered trademark of Merck KGaA, Darmstadt, Germany