Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
HPLC Analysis of Organo-acid Pesticides on Discovery® C8
HPLC Analysis of Organo-acid Pesticides on Discovery® C8
Conditions
column Discovery C8, 15 cm x 4.6 mm I.D., 5 µm particles (59353-U)
column temp. 20 °C
mobile phase [A] 25 mM potassium phosphate, (pH 2.3): [B] acetonitrile; (60:40, A:B)
flow rate 2 mL/min
injection 1 µL
detector UV, 214 nm
Description
Categories Analytical Chromatography, Environmental Analysis, Organics, Pesticides & Herbicides, Chromatograms, HPLC Chromatograms
Featured Industry Environmental
Legal Information Discovery is a registered trademark of Sigma-Aldrich Co. LLC