Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
HPLC Analysis of PA-Glycans from Mouse Monoclonal Antibody (MAb) on TSKgel® Amide-80, HILIC Mode
HPLC Analysis of PA-Glycans from Mouse Monoclonal Antibody (MAb) on TSKgel® Amide-80, HILIC Mode
Conditions
column TSKgel Amide-80, 25 cm x 4.6 mm I.D., 5 µm particles (813071)
column temp. 40 °C
mobile phase [A] 0.2 M acetic acid + TEA (pH 7.3); [B] acetonitrile
gradient 75% B for 5 min; to 50% B in 75 min
flow rate 0.5 mL/min
sample pyridylaminated oligosaccharides released from mouse mAb-1
injection 50 µL
detector fluorescence, 315/380, ex/em
Description
Categories Analytical Chromatography, Carbohydrates
Featured Industry Life Science and Biopharma
Legal Information TSKgel is a registered trademark of Tosoh Corporation
Other Notes Data provided by Tosoh Bioscience LLC.