Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
HPLC Analysis of Paraben Preservatives on Discovery® C8
HPLC Analysis of Paraben Preservatives on Discovery® C8
Conditions
column Discovery C8, 15 cm x 4.6 mm I.D., 5 µm particles (59353-U)
column temp. 20 °C
mobile phase [A] acetonitrile: [B] water, (30:70, A:B)
flow rate 1 mL/min
injection 10 µL
detector UV, 254 nm
Description
Categories Analytical Chromatography, Preservatives
Featured Industry Cosmetics, Personal Care, and Cleaning Products
Legal Information Discovery is a registered trademark of Sigma-Aldrich Co. LLC