Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
HPLC Analysis of Phenolic Compounds on Ascentis® Express 2.7 μm RP-Amide
HPLC Analysis of Phenolic Compounds on Ascentis® Express 2.7 μm RP-Amide
Conditions
column Ascentis Express RP-Amide, 10 cm x 4.6 mm I.D., 2.7 µm particles (53929-U)
column temp. 35 °C
mobile phase [A] 20 mM phosphoric acid, pH 2; [B] acetonitrile; (75:25, A:B)
flow rate 1.5 mL/min
sample 50 mg/L in mobile phase
injection 25 µL
detector UV, 270 nm
Description
Analysis Note Chromatography of Seven Phenol Compounds on Ascentis Express RPA
Categories Analytical Chromatography, Phenols
Featured Industry Vitamins, Nutraceuticals, and Natural Products