Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
HPLC Analysis of Phenytoin on Chromolith(R) Performance 250 - 4.6 mm
HPLC Analysis of Phenytoin on Chromolith® Performance 250 - 4.6 mm
Conditions
sample preparation Mobile phase
column Chromolith® Performance 250 x 4.6 mm (1.02129.0001)
column temp. ambient
mobile phase [A] metanol; [B] water; (50:50, A:B)
flow rate 0.5 mL/min
pressure 406 psi 28 Bar
sample 1 ppm of each Phenytoin & all related impurities
injection 10 μL
detector 229 nm cell: 10 μL
Description
Featured Industry Pharmaceutical (small molecule)
Legal Information Chromolith is a registered trademark of Merck KGaA, Darmstadt, Germany