HPLC Analysis of Pioglitazone Hydrochloride on Chromolith(R) HighResolution RP-18 endcapped 150 x 4.6mm
HPLC Analysis of Pioglitazone Hydrochloride on Chromolith® HighResolution RP-18 endcapped 150 x 4.6mm
Conditions
column Chromolith® HighResolution RP-18 endcapped, 150 x 4.6 mm (1.52023.0001)
column temp. ambient
mobile phase [A] acetonitrile; [B] 0.1 M ammonium acetate; [C] glacial acetic acid; (25:25:1, A:B:C))
flow rate 0.7 mL/min
pressure 972 psi (67 Bar)
sample 50 μg/mL of Pioglitazone HCl and 13 μg/mL of benzophenone (SST solution)
injection 20 μL
detector 269 nm cell: 13 μL
Description
Featured Industry Pharmaceutical (small molecule)
Legal Information Chromolith is a registered trademark of Merck KGaA, Darmstadt, Germany