HPLC Analysis of Polar and Non-Polar Analytes on Ascentis® Express C8, Column Test Mixture
HPLC Analysis of Polar and Non-Polar Analytes on Ascentis® Express C8, Column Test Mixture
Conditions
column Ascentis Express C8, 5 cm x 4.6 mm I.D., 2.7 µm particles (53512-U)
column temp. 30 °C
mobile phase [A] water; [B] acetonitrile; (45:55, A:B)
flow rate 1.76 mL/min
injection 5 µL
detector UV, 254 nm
Description
Categories Analytical Chromatography, Hydrocarbons