Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
HPLC Analysis of Polyphenols in Nero d'Avola Red Wine on Discovery® HS C18 (UV 354 nm)
HPLC Analysis of Polyphenols in Nero d'Avola Red Wine on Discovery® HS C18 (UV 354 nm)
Conditions
sample/matrix Nero d'Avola wine sample filtered prior injection through a 0.45 µm nylon membrane
column Discovery HS C18, 15 cm x 2.1 mm I.D., 3 µm particles (569255-U)
column temp. 30 °C
mobile phase [A] water, pH 3 with fomic acid; [B] Acetonitrile, pH 3 with fomic acid
flow rate 0.2 mL/min
injection 5 µL
detector UV, 354 nm
Description
Categories Analytical Chromatography, Food & Beverage Analysis, Flavors, Fragrances, Beverage Testing, Beer, Wine, and Spirits, Wine
Featured Industry Food and Beverages
Legal Information Discovery is a registered trademark of Sigma-Aldrich Co. LLC