Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
HPLC Analysis of Positional Isomers (Xylenes) on Astec® CYCLOBOND® I 2000
HPLC Analysis of Positional Isomers (Xylenes) on Astec® CYCLOBOND® I 2000
Conditions
column CYCLOBOND I 2000, 25 cm x 4.6 mm I.D., 5 µm particles (20024AST)
column temp. 55 °C
mobile phase [A] acetonitrile; [B] water; (15:85, A:B)
flow rate 0.8 mL/min
sample xylene isomers, each compound, 0.1 mg/mL in acetonitrile:water (50:50) ((elution order: m, o, p))
injection 3 µL
detector UV, 230 nm
Description
Categories Analytical Chromatography, Hydrocarbons
Featured Industry Chemicals and Industrial Polymers
Legal Information Astec is a registered trademark of Sigma-Aldrich Co. LLC, CYCLOBOND is a registered trademark of Sigma-Aldrich Co. LLC