Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
HPLC Analysis of Proteins on TSKgel® Phenyl-5PW
HPLC Analysis of Proteins on TSKgel® Phenyl-5PW
Conditions
column TSKgel Phenyl-5PW, 7.5 cm x 7.5 mm I.D., 10 mum particles (807573)
mobile phase 0.1 M phosphate buffer, pH 7.0 plus ammonium sulfate, 1.8 M to 0 M in 60 min
flow rate 1 mL/min
detector UV, 280 nm
Description
Categories Analytical Chromatography, Proteins
Featured Industry Life Science and Biopharma
Legal Information TSKgel is a registered trademark of Tosoh Corporation