Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
HPLC Analysis of Proteins on Superdex® Gel Filtration column
HPLC Analysis of Proteins on Superdex® Gel Filtration column
Conditions
column HiLoad 16/60 Superdex® 75 prep grade (54802-U)
mobile phase 0.15M ammonium acetate, pH 6.0
flow rate 0.75 mL/min
sample IGF-1, ZZ fusion protein and impurties
detector UV, 280 nm
Description
Categories Analytical Chromatography, Proteins
Featured Industry Life Science and Biopharma
Legal Information Superdex is a registered trademark of GE Healthcare