Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
HPLC Analysis of Proteins by Size Exclusion on Zenix® and SRT® SEC-300, Comparison of Particle Size and Column I.D.
HPLC Analysis of Proteins by Size Exclusion on Zenix® and SRT® SEC-300, Comparison of Particle Size and Column I.D.
Conditions
column SRT SEC-300, 30 cm x 7.8 mm 5 µm particles, 300 Ã… (Z777051)
column Zenix 300, 30 cm x 4.6 mm I.D., 3 µm particles, 300 Ã… (Z777028)
column Zenix 300, 30 cm x 7.8 mm I. D., 3 µm particles, 300 Ã… (Z777033)
column temp. ambient
mobile phase 150 mM sodium phosphate, pH 7
sample each compound 1 mg/mL except uracil, 2.5 µg/mL
detector UV, 214nm
Description
Categories Analytical Chromatography, Proteins
Featured Industry Life Science and Biopharma
Legal Information SRT is a registered trademark of Sepax Technologies, Zenix is a registered trademark of Sepax Technologies