Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
HPLC Analysis of Proteins Comparing Ion Selectivity on Proteomix® WCX-NP5
HPLC Analysis of Proteins Comparing Ion Selectivity on Proteomix® WCX-NP5
Conditions
column Proteomix WCX-NP5, 5 cm x 4.6 mm I.D., 5 µm particles (Z777193)
column temp. 25 °C
mobile phase [A] 10 mM potassium phosphate, dibasic, pH 7 with phosphoric acid; [B] 0.3 M potassium potassium phosphate, pH 7 with phosphoric acid; ([C] 10 mM sodium phosphate, dibasic, pH 7 with phosphoric acid; [D] 0.3 M sodium potassium phosphate, dibasic, pH 7 with phosphoric acid;)
gradient 100% A to 100% B (or 100% C to 100% D) in 20 min
flow rate 0.5 mL/min
pressure 615 psitotal pressure (42 bar total pressure)
sample g/L ea: ribonuclease a, 0.38; cytochrome c, 0.15; lysozyme, 0.23; a-chymptrypsinogen, 0.23
injection 5 µL
detector UV, 280 nm
Description
Analysis Note The choice of exchangable counterion can affect retention and selectivity with ion exchange chromatography of proteins. In this case, under the chromatographic conditions employed, differences are subtle.
Categories Analytical Chromatography, Proteins
Featured Industry Life Science and Biopharma
Legal Information Proteomix is a registered trademark of Sepax Technologies