Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
HPLC Analysis of Proteins on Discovery® BIO Wide Pore C5
HPLC Analysis of Proteins on Discovery® BIO Wide Pore C5
Conditions
column Discovery BIO Wide Pore C5, 15 cm x 4.6 mm I.D., 5 µm particles (568422-U)
column temp. ambient
mobile phase [A] (0.1% TFA in water): [B] (0.1% TFA in CH3CN); (75:25, A:B)
gradient 0-100% B in 25 min
flow rate 1 mL/min
injection 12 µLin 0.1% TFA
detector UV, 220 nm
Description
Categories Analytical Chromatography, Proteins
Featured Industry Life Science and Biopharma
Legal Information Discovery is a registered trademark of Sigma-Aldrich Co. LLC