Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
HPLC Analysis of Proteins on TSKgel® BioAssist S, Ion Selectivity Comparison
HPLC Analysis of Proteins on TSKgel® BioAssist S, Ion Selectivity Comparison
Conditions
column TSKgel BioAssist S, 5 cm x 4.6 mm I.D., 7 µm particles (819686)
column temp. 25 °C
mobile phase [A] 10 mM potassium phosphate, dibasic, pH 7 with phosphoric acid; [B] 0.3 M potassium potassium phosphate, pH 7 with phosphoric acid; ([C] 10 mM sodium phosphate, dibasic, pH 7 with phosphoric acid; [D] 0.3 M sodium potassium phosphate, dibasic, pH 7 with phosphoric acid;)
gradient 100% A to 100% B (or 100% C to 100% D) in 33 min, held at 100% B (or D) for 5 minutes
flow rate 0.3 mL/min
pressure 615 psitotal pressure (42 bar total pressure)
sample g/L ea: ribonuclease a, 0.38; cytochrome c, 0.15; lysozyme, 0.23; a-chymptrypsinogen, 0.23
injection 5 µL
detector UV, 280 nm
Description
Analysis Note The choice of exchangable counterion can affect retention and selectivity when performing ion exchange chromatography of proteins. This application note compares sodium and potassium gradients for changes in retention and selectivity.
Categories Analytical Chromatography, Proteins
Featured Industry Life Science and Biopharma
Legal Information TSKgel is a registered trademark of Tosoh Corporation