HPLC Analysis of PVP-15 Polymer on TSKgel® SuperMultiporePW-N
HPLC Analysis of PVP-15 Polymer on TSKgel® SuperµLtiporePW-N
column TSKgel SuperµLtiporePW-N, 15 cm x 6 mm I.D., 4 µm particles (822789)
column temp. 40 °C
mobile phase 0.1 M sodium nitrate
flow rate 0.6 mL/min
sample PVP-15 polymer
injection 20 µL
detector refractive index (RI)
Analysis Note Hydrophobic interaction causes partial adsorption of PVP-15 polymer on TSKgel G3000PWxl and G2500PWxl columns, while the absence of adsorption on the TSKgel SuperµLtiporePW-N column suggests that the particle surface is more hydrophilic than the conventional columns.
Categories Analytical Chromatography, Polymers, coatings, and plastics
Featured Industry Chemicals and Industrial Polymers
Legal Information TSKgel is a registered trademark of Tosoh Corporation
Other Notes Data provided by Tosoh Bioscience LLC.