Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
HPLC Analysis of Quaternary Ammonium Drugs on Discovery® HS F5
HPLC Analysis of Quaternary Ammonium Drugs on Discovery® HS F5
Conditions
column Discovery HS F5, 15 cm x 4.6 mm I.D., 5 µm particles (567516-U)
column temp. 60 °C
mobile phase 10 mM ammonium acetate in 80% acetonitrile
flow rate 1 mL/min
sample 50 mg/mL in 70:30, 10 mM ammonium acetate in 80% acetonitrile:methanol
injection 10 µL
detector UV, 225 nm
Description
Categories Analytical Chromatography, Pharmaceutical Analysis, Anesthetic Drugs
Featured Industry Pharmaceutical (small molecule)
Legal Information Discovery is a registered trademark of Sigma-Aldrich Co. LLC