HPLC Analysis of Ranitidine on Chromolith(R) High Resolution RP-18e
HPLC Analysis of Ranitidine on Chromolith® High Resolution RP-18e
Conditions
column Chromolith® High Resolution RP-18 endcapped, 100 x 4.6 mm (1.52022.0001)
column temp. 30 °C
mobile phase [A] acetonitrile; [B] 10mM Phosphat buffer pH=7; (5:95, A:B)
flow rate 1.0 mL/min
sample Ranitidine 0,15mg/mL (Diluent: solved in mobile phase)
injection 5 μL
detector 320 nm cell: 16 μL
Description
Featured Industry Pharmaceutical (small molecule)
Legal Information Chromolith is a registered trademark of Merck KGaA, Darmstadt, Germany