Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
HPLC Analysis of Sigma Protein Standard Mix on Zenix® SEC-300
HPLC Analysis of Sigma Protein Standard Mix on Zenix® SEC-300
Conditions
column Sepax Zenix SEC-300, 30 cm x 4.6 mm I.D., 3 µm particles (Z777028)
column temp. 25 °C
mobile phase 0.15 M potassium phosphate monobasic, pH 7.0 with potassium hydroxide
flow rate 0.3 mL/min
pressure 1050 psi (72 bar)
sample 0.5 mL of mobile phase added to one vial of  Sigma Protein Standard Mix   (p/n 69385); filtered 0.45 µm Millex-HV syringe filter (p/n Z227552)
injection 2.5 µL
detector UV, 215 & 280 nm
Description
Featured Industry Life Science and Biopharma
Legal Information Zenix is a registered trademark of Sepax Technologies