HPLC Analysis of Sildenafil and Impurities on Chromolith(R) Performance (4.6 mm I.D. columns)
HPLC Analysis of Sildenafil and Impurities on Chromolith® Performance (4.6 mm I.D. columns)
Conditions
column Chromolith® Performance RP-18 endcapped, 10 cm x 4.6 mm I.D. (2 columns in series) (1.02129.0001)
column temp. ambient
mobile phase A] acetonitrile; [B] na-phosphatbuffer 20mM pH=3.0;
gradient Initial composition: 0-8min 55%A to 45%B isocratic, flow increases from 0.5 to 1mL/min, 8,1-20min 65%A to 35%B isocratic, flow 3mL/min.
pressure 1450 psi (100 bar)
sample Methoxyamin (1mg/ml), Sildenafil Citrat (5mg/ml), Amidin (1mg/ml), Imidon 1 (0.5mg/ml), Imidon 2 (0.5mg/ml), Thiophenethylester (1mg/ml), Acyl-BT-Ester (2mg/ml),Aminacid (0,5mg/ml), CHT-Ester (1mg/ml), Acyl-CHT-Ester (2mg/ml),Aminester (0.2mg/ml), Imidochlorid 1 (0.3mg/ml), Imidochlorid 2 (0.2mg/ml)
injection 0.5 μL
detector 254 nm cell: 11 μL
Description
Featured Industry Pharmaceutical (small molecule)
Legal Information Chromolith is a registered trademark of Merck KGaA, Darmstadt, Germany