Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
HPLC Analysis of Sildenafil Citrate and Related Substances on Chromolith High Resolution RP-18e
HPLC Analysis of Sildenafil Citrate and Related Substances on Chromolith High Resolution RP-18e
Conditions
column Chromolith High Resolution RP-18 endcapped, 100 x 4.6 mm (1.52022.0001)
column temp. 30 °C
mobile phase [A] 7 mL of triethylamine with water to 1mL. Adjust with phosphoric acid to a pH=3.0±0.1; [B] methanol; [C] acetonitrile; (58:25:17, A:B:C)
flow rate 1.0 mL/min
pressure 971.5 psi (67 bar)
sample Sildenafil Citrate RS 28ppm, Sildenafil impurity A 28ppm .
injection 20 μL
detector 290 nm (cell: 13 μL)