HPLC Analysis of Sildenafil Citrate on Chromolith(R) HighResolution RP-18 endcapped 100 x 4.6mm
HPLC Analysis of Sildenafil Citrate on Chromolith® HighResolution RP-18 endcapped 100 x 4.6mm
Conditions
column Chromolith HighResolution RP-18 endcapped, 100 x 4.6 mm (1.52022.0001)
column temp. 30 °C
mobile phase [A] Dilute 7 mL of triethylamine with water to 1mL. Adjust with phosphoric acid to a pH=3.0±0.1; [B] methanol; [C] acetonitrile; (58:25:17, A:B:C)
flow rate 1.0 mL/min
pressure 971.5 psi (67 bar)
sample Sildenafil Citrate RS 28 ppm, Sildenafil Related Substance A 28ppm
injection 20 μL
detector 290 nm cell: 13 μL
Description
Featured Industry Pharmaceutical (small molecule)
Legal Information Chromolith is a registered trademark of Merck KGaA, Darmstadt, Germany