Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
HPLC Analysis of Simvastatine USP on Chromolith(R) Performance Flash 25-4.6mm
HPLC Analysis of Simvastatine USP on Chromolith® Performance Flash 25-4.6mm
Conditions
sample preparation 1.4mg/ml monobasic potassium phosphate in water
column Chromolith Performance Flash 25-4.6mm (1.51463.0001)
column temp. ambient
mobile phase [A] 50% Acetonitril, 50% phosphoric acid and water 1:999; [B] 100% phosphoric acid and Acetonitril 1:999;
gradient Initial composition:0-4.5min 100%A, 4.6min to 8 min 95%A to 25%A (8min to 11.5min 25%A, 11.6min to 13min 100%A)
flow rate 3 mL/min
pressure 8337 psi (37-58 bar)
sample Lovastatin (1,5mg/ml), Simvastatin (1,5mg/ml) (Diluent: Acetonitril and Buffer 3:2)
injection 5 μL
detector 238 nm (cell: 11 μL)
Description
Featured Industry Pharmaceutical (small molecule)
Legal Information Chromolith is a registered trademark of Merck KGaA, Darmstadt, Germany