Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
HPLC Analysis of Substituted Aromatic Compounds on Ascentis® Express Biphenyl
HPLC Analysis of Substituted Aromatic Compounds on Ascentis® Express Biphenyl
Conditions
column Ascentis Express Biphenyl, 10 cm x 2.1 mm, 2.7 µm particles (64065-U)
column temp. 35 °C
mobile phase [A] Water; [B]Methanol; (57:43 A:B)
flow rate 0.5 mL/min
pressure 7900 psi (545 bar)
sample Uracil (25 µg/mL), Benzonitrile (50 µg/mL), Acetophenone (25 µg/mL), Anisole (100 µg/mL),
injection 1 µL
detector UV, 254 nm
Description
Analysis Note This application demonstrate the separation of substituted aromatic compounds on Ascentis Express Biphenyl. These analytes highlight the unique selectivity of this stationary phase for this type of analytes using methanol reversed phase conditions.
Categories Analytical Chromatography, Aromatics
Featured Industry Chemicals and Industrial Polymers
General Analytical