Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
HPLC Analysis of Sugars and Sugar Enantiomers on Astec® CYCLOBOND® I 2000
HPLC Analysis of Sugars and Sugar Enantiomers on Astec® CYCLOBOND® I 2000
Conditions
column CYCLOBOND I 2000, 25 cm x 4.6 mm I.D., 5 µm particles (20024AST)
column temp. 35 °C
mobile phase [A] water; [B] acetonitrile; (15:85, A:B)
flow rate 1 mL/min
sample 0.5 mg/mL each in water:acetonitrile, 25:75
injection 10 µL
detector ELSD at 35C and 3.2 bar nitrogen
Description
Categories Analytical Chromatography, Chiral Applications, Carbohydrates, Food & Beverage, Life Science & Biopharma, All Biochemical Applications, All Food & Beverage Applications
Featured Industry Food and Beverages
Legal Information Astec is a registered trademark of Sigma-Aldrich Co. LLC, CYCLOBOND is a registered trademark of Sigma-Aldrich Co. LLC