Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
HPLC Analysis of Tartaric Acid Enantiomers on Astec® CLC-D, Trace Level Detection
HPLC Analysis of Tartaric Acid Enantiomers on Astec® CLC-D, Trace Level Detection
Conditions
column Astec CLC-D, 15 cm x 4.6 mm I.D., 5 µm particles (53023AST)
column temp. 25 °C
mobile phase 3 mM copper sulfate, pH 3.2
flow rate 1 mL/min
sample 5 mg/mL in water
injection 10 µL
detector UV, 254 nm
Description
Categories Analytical Chromatography, Chiral Applications, Food & Beverage, Organic Acids, Chemicals and Industrial Polymers, All Food & Beverage Applications
Featured Industry Chemicals and Industrial Polymers
Food and Beverages
Vitamins, Nutraceuticals, and Natural Products
Legal Information Astec is a registered trademark of Sigma-Aldrich Co. LLC