HPLC Analysis of Toluene, Uracil and Cytosine on Chromolith(R) Performance Diol 100-4.6mm
HPLC Analysis of Toluene, Uracil and Cytosine on Chromolith® Performance Diol 100-4.6mm
Conditions
column Chromolith® Performance DIOL, 100 x 4.6 mm (1.53172.0001)
column temp. 23 °C
mobile phase [A] acetonitrile; [B] 5mM Ammoniumacetate buffer pH=6.8; (80:20, A:B)
flow rate 0.5 mL/min
pressure 87 psi (6 Bar)
sample Toluene 0,67mg/ml, Uracil 0,04mg/ml, Cytosine 0,06mg/ml (Diluent: Mobile phase)
injection 10 μL
detector 254 nm
Description
Featured Industry General Analytical
Legal Information Chromolith is a registered trademark of Merck KGaA, Darmstadt, Germany