HPLC Analysis of Tolvaptan on Chromolith(R) High Resolution RP-18e
HPLC Analysis of Tolvaptan on Chromolith® High Resolution RP-18e
column Chromolith® High Resolution RP-18 endcapped, 100 x 4.6 mm (1.52022.0001)
column temp. 40 °C
mobile phase [A] 1.15 g of ammonium dihydrogen phosphate & 1.32 gm of di-ammonium hydrogen phosphate 1L Milli-Q® water. (pH is about 6.8); [B] acetonitrile; (75:25, A:B)
flow rate 1.5 mL/min
sample Weigh 25 mg substance in 25 ml volumetric flask. Dissove in diluent & make up the volume with diluent.
injection 10 μL
detector 265 nm cell: 10 μL
Featured Industry Pharmaceutical (small molecule)
Legal Information Chromolith is a registered trademark of Merck KGaA, Darmstadt, Germany