HPLC Analysis of Travoprost on Chromolith(R) High Resolution RP-18e
HPLC Analysis of Travoprost on Chromolith® High Resolution RP-18e
Conditions
column Chromolith High Resolution RP-18 endcapped, 100 x 4.6 m (1.52022.0001)
column temp. ambient
mobile phase [A] acetonitrile; [B] water + 0,1% TFA v/v; (80:20, A:B)
flow rate 1.0 mL/min
sample Travoprost (Fluprostenol Isopropylacetat) (1mg/mL), Travoprost free acid (Fluprostenol) (1mg/mL)
injection 0.5 μL
detector 210 nm cell: 11 μL
Description
Featured Industry Pharmaceutical (small molecule)
Legal Information Chromolith is a registered trademark of Merck KGaA, Darmstadt, Germany