HPLC Analysis of Travoprost on Chromolith(R) Performance 100-4.6 mm
HPLC Analysis of Travoprost on Chromolith® Performance 100-4.6 mm
Conditions
column Chromolith® Performance, 100 x 4.6 mm (1.52028.0001)
column temp. ambient
mobile phase [A] acetonitrile; [B] water plus 0,1% TFA v/v; (80:20, A:B)
flow rate 1 mL/min
pressure 4744 psi (327 bar)
sample Travoprost (Fluprostenol Isopropylacetat) (1mg/mL), Travoprost free acid (Fluprostenol) (1mg/mL)
injection 1 μL
detector 210 nm cell: 11 μL
Description
Featured Industry Pharmaceutical (small molecule)
Legal Information Chromolith is a registered trademark of Merck KGaA, Darmstadt, Germany