HPLC Analysis of Travoprost on Chromolith(R) Performance 100-3mm
HPLC Analysis of Travoprost on Chromolith® Performance 100-3mm
Conditions
column Chromolith® Performance, 100 x 3 mm (1.52001.0001)
column temp. ambient
mobile phase [A] acetonitrile; [B] water + 0,1% TFA v/v; (80:20, A:B)
flow rate 425 μL/min
pressure 4457 psi (307 bar)
sample Travoprost (Fluprostenol Isopropylacetat) (1mg/mL), Travoprost free acid (Fluprostenol) (1mg/mL)
injection 0.2 μL
detector 210 nm (cell: 1.4 μL)
Description
Featured Industry Pharmaceutical (small molecule)
Legal Information Chromolith is a registered trademark of Merck KGaA, Darmstadt, Germany