Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
HPLC Analysis of Tricyclic Antidepressants on Discovery® Cyano
HPLC Analysis of Tricyclic Antidepressants on Discovery® Cyano
Conditions
column Discovery Cyano, 15 cm x 4.6 mm I.D., 5 µm particles (59356-U)
column temp. 35 °C
mobile phase [A] acetonitrile: [B] methanol: [C] 10 mM potassium phosphate, dibasic, pH 7.0, (60:20:20, A:B:C)
flow rate 1 mL/min
detector UV, 254 nm
Description
Categories Analytical Chromatography, Pharmaceutical Analysis, Antidepressants
Featured Industry Pharmaceutical (small molecule)
Legal Information Discovery is a registered trademark of Sigma-Aldrich Co. LLC