Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
HPLC Analysis of Tricyclic Antidepressants on Ascentis® C8
HPLC Analysis of Tricyclic Antidepressants on Ascentis® C8
Conditions
column Ascentis C8, 15 cm x 2.1 mm I.D., 3 µm particles (581402-U)
column temp. 35 °C
mobile phase [A] 25 mM ammonium phosphate dibasic (pH 7.0 with phosphoric acid): [B] methanol; (25:75, A:B)
flow rate 0.2 mL/min
sample as indicated in 60:40 water:methanol
injection 10 µL
detector UV, 254 nm
Description
Categories Analytical Chromatography, Clinical, Pharmaceutical Analysis, antidepressants, Antidepressants
Featured Industry Clinical
Pharmaceutical (small molecule)