Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
HPLC Analysis of Tryptic Digest on BIOshell™ A160 Peptide C18
HPLC Analysis of Tryptic Digest on BIOshell™ A160 Peptide C18
Conditions
column BIOshell A160 Peptide C18, 15 cm x 2.1 mm I.D., 2.7 µm particles (66905-U)
column temp. 35 °C
mobile phase [A] 0.1% formic acid in water; [B] 25:75, (0.4% formic acid in water):acetonitrile
gradient 0 to 70% B in 40 min.
flow rate 0.3 mL/min
sample 10 pg/L in 0.1% formic acid
injection 2 µL
detector ESI(+), TOF
Description
Categories Analytical Chromatography, Proteins
Featured Industry Life Science and Biopharma
Legal Information BIOshell is a trademark of Sigma-Aldrich Co. LLC