Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
HPLC Analysis of Ulcer Treatment Drugs on Ascentis® RP-Amide
HPLC Analysis of Ulcer Treatment Drugs on Ascentis® RP-Amide
Conditions
column Ascentis RP-Amide, 15 cm x 2.1 mm I.D., 5 µm particles (565305-U)
column temp. 35 °C
mobile phase [A] 10 mM ammonium phosphate, pH 6.2 with phosphoric acid; [B] acetonitrile
gradient 5 to 15% B in 10 min; to 55% B in 8 min; held for 5 min
flow rate 0.2 mL/min
sample as indicated in 95:5 water:methanol
injection 5 µL
detector UV, 220 nm
Description
Categories Analytical Chromatography, Antiulcer Compounds