HPLC Analysis of UMP, UDP and UTP on SeQuant(R) ZIC-HILIC
HPLC Analysis of UMP, UDP and UTP on SeQuant® ZIC-HILIC
Conditions
column SeQuant ZIC-HILIC, PEEK, 100 x 4.6 mm, 5 μm, 200 Ã… (1.50453.0001)
mobile phase [A] acetonitrile; [B] ammonium acetate; (70:30, A,B)
flow rate 0.7 mL/min
injection 5 μL
detector 259 nm
Description
Featured Industry Pharmaceutical (small molecule)
Legal Information SeQuant is a registered trademark of Merck KGaA, Darmstadt, Germany