Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
HPLC Analysis of Various Cationic Polymers on TSKgel® PWXL-CP Columns
HPLC Analysis of Various Cationic Polymers on TSKgel® PWXL-CP Columns
Conditions
column TSKgel G3000PWXL-CP, 30 cm x 7.8 mm, 7 µm particles (821873)
column TSKgel G6000PWXL-CP, 30 cm x 7.8 mm, 13 µm particles (821875)
column TSKgel G5000PWXL-CP, 30 cm x 7.8 mm, 10 µm particles (821874)
column temp. 25 °C
mobile phase 0.1 M sodium nitrate
flow rate 1 mL/min
sample 3 mg/mL
injection 100 µL
detector refractive index (RI)
Description
Analysis Note Various cationic polymers with different functional groups and molecular weights were injected on the TSKgel PWXL-CP columns (TSKgel G6000PWXL-CP, G5000PWXL-CP and G3000PWXL-CP, connected in series).
Categories Analytical Chromatography, Polymers, coatings, and plastics
Featured Industry Chemicals and Industrial Polymers
Legal Information TSKgel is a registered trademark of Tosoh Corporation
Other Notes Data provided by Tosoh Bioscience LLC.