HPLC Analysis of Vitamin E on Chromolith(R) High Resolution RP-18e
HPLC Analysis of Vitamin E on Chromolith® High Resolution RP-18e
Conditions
column Chromolith® High Resolution RP-18 encapped, 100 x 4.6 mm (1.52022.0001)
column temp. 25 °C
mobile phase [A] methanol; [B] acetonitrile; (92:8, A:B)
flow rate 1.0 mL/min
sample 1000 ppm sample in mobile phase
injection 5 μL
detector 230 nm cell: 10μL
Description
Featured Industry Food and Beverages
Legal Information Chromolith is a registered trademark of Merck KGaA, Darmstadt, Germany