Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
HPLC Analysis of Vitamins, Fat Soluble (A and E), on SUPELCOSIL(TM) LC-18
HPLC Analysis of Vitamins, Fat Soluble (A and E), on SUPELCOSIL™ LC-18
Conditions
column SUPELCOSIL LC-18, 25 cm × 4.6 mm I.D., 5 μm particles (58298)
mobile phase [A] methanol: [B] deionized water (98:2, A:B)
flow rate 2 mL/min
sample 0.2-1 mg/mL each analyte in methanol
injection 20 μL
detector UV, 325 nm (retinol, retinyl acetate) or 290 nm (tocopherols, tocopherol acetate)
Description
Categories Analytical Chromatography, Vitamins and Natural products
Featured Industry Clinical
Vitamins, Nutraceuticals, and Natural Products
Legal Information SUPELCOSIL is a trademark of Sigma-Aldrich Co. LLC