HPLC Analysis of Voriconazole With Acidic Stress on Chromolith Performance 100-3mm
HPLC Analysis of Voriconazole With Acidic Stress on Chromolith Performance 100-3mm
Conditions
sample preparation stirring 1 hour in HCl 0.1N
column Chromolith® Performance 100 x 3 mm (1.52001.0001)
column temp. ambient
mobile phase [A] acetonitrile; [B] water; (50:50, A:B)
flow rate 425 μL/min
pressure 4032 psi (28 bar)
sample Voriconazole 50 μg/mL
injection 5 μL
detector 256 nm (cell: 1.4 μL)