Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
HPLC Analysis of Voriconazole With Exposition To UV Light on Chromolith Performance 100-4.6 mm
HPLC Analysis of Voriconazole With Exposition To UV Light on Chromolith Performance 100-4.6 mm
Conditions
sample preparation stirring 3 hours under UV-Light 254nm
column Chromolith® Performance 100 x 4.6 mm (1.52028.0001)
column temp. ambient
mobile phase [A] acetonitrile; [B] water; (50:50, A:B)
flow rate 1 mL/min
pressure 4758 psi (33 bar)
sample Voriconazole 50 μg/mL
injection 20 μL
detector 256 nm cell: 11 μL