HPLC Analysis of Voriconazole With Heat Stress on Chromolith Performance 100-2mm
HPLC Analysis of Voriconazole With Heat Stress on Chromolith Performance 100-2mm
Conditions
sample preparation stirring 24 hours at 70°C
column Chromolith® Performance 100 x 2 mm (1.52006.0001)
column temp. ambient
mobile phase [A] acetonitrile; [B] water; (50:50, A:B)
flow rate 190 μL/min
pressure 302 psi (21 bar)
sample Voriconazole 50 μg/mL
injection 2 μL
detector 256 nm cell: 1.4 μL