HPLC Analysis of Voriconazole With Oxidation on Chromolith Performance 100-4.6 mm
HPLC Analysis of Voriconazole With Oxidation on Chromolith Performance 100-4.6 mm
Conditions
column Chromolith® Performance 100 x 4.6 mm (1.52028.0001)
column temp. ambient
mobile phase [A] acetonitrile; [B] water; (50:50, A:B)
flow rate 1 mL/min
pressure 4758 psi (33 bar)
sample Voriconazole 50 μg/mL
injection 20 μL
detector 256 nm cell: 11 μL